4H-Furo[3,2-b]pyrrole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A716670-100MG A716670-1G A716670-250MG A716670-5G
77293-384EA 65.92 USD
77293-384 77293-386 77293-388 77617-946
4H-Furo[3,2-b]pyrrole-5-carboxylic acid 98%
4H-Furo[3,2-b]pyrrole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₃
MDL Number: MFCD07368660
CAS Number: 67268-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR