1,4-Di(4H-1,2,4-triazol-4-yl)benzene 97%
Supplier: Ambeed
|
Warning
|
A716583-1G
A716583-250MG
A716583-5G
77267-666EA
173.04
USD
77267-666
77267-668
77267-670
1,4-Di(4H-1,2,4-triazol-4-yl)benzene 97%
1,4-Di(4H-1,2,4-triazol-4-yl)benzene
Precaución: for Research Use Only
Formula:
C₁₀H₈N₆ MW: 212.21 g/mol |
MDL Number:
MFCD31380110 CAS Number: 681004-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...