2-Iodo-3-methylbenzamide 97%

Supplier: Ambeed
Warning

A716413-1G A716413-250MG A716413-5G
77267-660EA 78.28 USD
77267-660 77267-662 77267-664
2-Iodo-3-methylbenzamide 97%
2-Iodo-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈INO
MW: 261.05 g/mol
MDL Number: MFCD09753716
CAS Number: 887234-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR