Isopropyl ethanesulfonate 97%

Supplier: Ambeed
Warning

A716212-25G A716212-250MG
77267-658EA 78 USD
77267-658 77749-463
Isopropyl ethanesulfonate 97%
Isopropyl ethanesulfonate

Precaución: for Research Use Only

Formula: C₅H₁₂O₃S
MDL Number: MFCD01728622
CAS Number: 14245-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR