4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A715233-100MG A715233-1G A715233-250MG
77243-614EA 38.11 USD
77243-614 77243-616 77243-618
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂·ClH
MW: 249.693 g/mol
MDL Number: MFCD08276229
CAS Number: 41567-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR