(4-Fluoro-3,5-dimethylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-fluorophenylboronic acid

A714984-100MG A714984-250MG A714984-1G
77255-286EA 28.84 USD
77255-286 77255-288 77747-419
(4-Fluoro-3,5-dimethylphenyl)boronic acid 98%
(4-Fluoro-3,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₂
MW: 167.97 g/mol
MDL Number: MFCD08706256
CAS Number: 342636-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR