4,4'-(Ethyne-1,2-diyl)dibenzaldehyde 98%

Supplier: Ambeed
Warning

A714394-100MG A714394-1G
77267-654EA 23.69 USD
77267-654 77267-656
4,4'-(Ethyne-1,2-diyl)dibenzaldehyde 98%
4,4'-(Ethyne-1,2-diyl)dibenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₀O₂
MW: 234.24 g/mol
MDL Number: MFCD01573051
CAS Number: 84907-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR