2,3-Dihydrobenzofuran-5-acetic acid 97%

Supplier: Ambeed
Warning

A714320-100G A714320-10G A714320-1G A714320-25G A714320-5G
77255-264EA 143 USD
77255-264 77255-266 77255-268 77255-270 77255-272
2,3-Dihydrobenzofuran-5-acetic acid 97%
2,3-Dihydrobenzofuran-5-acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD01941301
CAS Number: 69999-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR