3-Chloro-1-phenyl-1-propyne 98%

Supplier: Ambeed
Warning

A714284-100MG A714284-1G A714284-250MG A714284-5G
77243-596EA 6 USD
77243-596 77243-598 77243-600 77243-602
3-Chloro-1-phenyl-1-propyne 98%
3-Chloro-1-phenyl-1-propyne

Precaución: for Research Use Only

Formula: C₉H₇Cl
MDL Number: MFCD06411085
CAS Number: 3355-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR