(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane 98%

Supplier: Ambeed
Warning

Synonyms: (R,R)-BDPP, (2R,4R)-Pentane-2,4-diylbis(diphenylphosphine)

A713973-100MG A713973-1G A713973-250MG A713973-5G
77255-256EA 41.2 USD
77255-256 77255-258 77255-260 77255-262
(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane 98%
(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane

Precaución: for Research Use Only

Formula: C₂₉H₃₀P₂
MW: 440.4960 g/mol
MDL Number: MFCD00142324
CAS Number: 96183-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR