1,4-Bis(4-aminophenoxy)butane 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(Butane-1,4-diylbis(oxy))dianiline, 4,4'-(1,4-Butanediyl)dioxydianiline

A713922-1G A713922-250MG
77293-364EA 26.78 USD
77293-364 77293-366
1,4-Bis(4-aminophenoxy)butane 97%
1,4-Bis(4-aminophenoxy)butane

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.3422 g/mol
MDL Number: MFCD00088690
CAS Number: 6245-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR