4-Phenoxybenzonitrile 98%
Supplier: Ambeed
|
Warning
|
A713736-100G
A713736-1G
A713736-25G
A713736-5G
77293-356EA
866.23
USD
77293-356
77293-358
77293-360
77293-362
4-Phenoxybenzonitrile 98%
4-Phenoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₁₃H₉NO MW: 195.22 g/mol |
MDL Number:
MFCD00017346 CAS Number: 3096-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...