2,3,5-Trifluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A713583-100G A713583-1G A713583-25G A713583-500G A713583-5G
77293-346EA 630.36 USD
77293-346 77293-348 77293-350 77293-352 77293-354
2,3,5-Trifluorophenylboronic acid 98%
2,3,5-Trifluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
MDL Number: MFCD01863166
CAS Number: 247564-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR