Lithium bis(fluorosulfonyl)imide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
LiFSI, Lithium bis(fluorosulphonyl)amide
A713428-25G
A713428-5G
77243-592EA
15
USD
77243-592
77243-594
Lithium bis(fluorosulfonyl)imide 98%
Lithium bis(fluorosulfonyl)imide
Precaución: for Research Use Only
Formula:
F₂LiNO₄S₂ |
MDL Number:
MFCD28126786 CAS Number: 171611-11-3 UN: 1759 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...