PBTZ169 97%

Supplier: Ambeed
Warning

Synonyms: Macozinone, 2-(4-(cyclohexylmethyl)piperazin-1-yl)-8-nitro-6-(trifluoromethyl)-4H-benzo[e][1,3]thiazin-4-one

A713021-10MG A713021-1MG A713021-25MG A713021-5MG
77243-584EA 55.62 USD
77243-584 77243-586 77243-588 77243-590
PBTZ169 97%
PBTZ169

Precaución: for Research Use Only

Formula: C₂₀H₂₃F₃N₄O₃S
MW: 456.48 g/mol
MDL Number: MFCD28902203
CAS Number: 1377239-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR