4-Methylnicotinamide 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-pyridinecarboxamide

A712989-100MG A712989-1G A712989-250MG
77293-340EA 32.96 USD
77293-340 77293-342 77293-344
4-Methylnicotinamide 95%
4-Methylnicotinamide

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.1512 g/mol
MDL Number: MFCD00234322
CAS Number: 7250-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR