(S)-Benzyl 2-(8-amino-1-bromoimidazo[1,5-a]pyrazin-3-yl)pyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A712917-100MG A712917-1G A712917-250MG A712917-5G
77255-248EA 22 USD
77255-248 77255-250 77255-252 77255-254
(S)-Benzyl 2-(8-amino-1-bromoimidazo[1,5-a]pyrazin-3-yl)pyrrolidine-1-carboxylate 97%
(S)-Benzyl 2-(8-amino-1-bromoimidazo[1,5-a]pyrazin-3-yl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₁₈BrN₅O₂
MDL Number: MFCD30184899
CAS Number: 1420478-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR