Chloro[(tricyclohexylphosphine)-2-(2'-aminobiphenyl)]palladium(II) 98%
Supplier: Ambeed
|
Warning
|
A712706-100MG
A712706-1G
A712706-250MG
A712706-5G
77255-236EA
35.02
USD
77255-236
77255-238
77255-240
77573-310
Chloro[(tricyclohexylphosphine)-2-(2'-aminobiphenyl)]palladium(II) 98%
Chloro[(tricyclohexylphosphine)-2-(2'-aminobiphenyl)]palladium(II)
Precaución: for Research Use Only
Formula:
C₃₀H₄₃ClNPPd MW: 590.52 g/mol |
MDL Number:
MFCD21363051 CAS Number: 1353658-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...