4-Methyl-2-thiazolecarboxaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 2-THIAZOLECARBOXALDEHYDE, 4-METHYL-, 4-Methylthiazole-2-carbaldehyde, 4-Methyl-1,3-thiazole-2-carboxaldehyde

A711366-250MG A711366-25G A711366-1G A711366-5G
77293-330EA 16.48 USD
77293-330 77293-332 77617-930 77749-453
4-Methyl-2-thiazolecarboxaldehyde 98%
4-Methyl-2-thiazolecarboxaldehyde

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.161 g/mol
MDL Number: MFCD00022450
CAS Number: 13750-68-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR