Ru(bpy)3(BF4)2 98%

Supplier: Ambeed
Warning

A711088-250MG A711088-1G
77280-906EA 76 USD
77280-906 77749-452
Ru(bpy)3(BF4)2 98%
Ru(bpy)3(BF4)2

Precaución: for Research Use Only

Formula: C₃₀H₂₄BF₄N₆Ru
MW: 656.42 g/mol
CAS Number: 63950-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR