S-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate 98%
Supplier: Ambeed
|
Warning
|
A710943-100MG
A710943-250MG
77255-224EA
94.76
USD
77255-224
77255-226
S-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate 98%
S-3,3'-Dibromo-1,1'-binaphthyl-2,2'-diylhydrogenphosphate
Precaución: for Research Use Only
Formula:
C₂₀H₁₁Br₂O₄P MW: 506.08 g/mol |
CAS Number:
1644121-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...