4,4'-Methylenedi-o-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 4,4'-Methylenebis(2-methylaniline)

A710898-10G A710898-25G A710898-5G
77293-324EA 30.9 USD
77293-324 77293-326 77293-328
4,4'-Methylenedi-o-toluidine 98%
4,4'-Methylenedi-o-toluidine

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂
MW: 226.3200 g/mol
MDL Number: MFCD00126963
CAS Number: 838-88-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR