Propan-2-yl 4-[5-methoxy-6-[(2-methyl-6-methylsulfonylpyridin-3-yl)amino]pyrimidin-4-yl]oxypiperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: APD-597, Isopropyl 4-((5-methoxy-6-((2-methyl-6-(methylsulphonyl)pyridin-3-yl)amino)pyrimidin-4-yl)oxy)piperidine-1-carboxylate

A710863-10MG A710863-1MG A710863-25MG A710863-50MG A710863-5MG
77243-572EA 180.25 USD
77243-572 77243-574 77243-576 77243-578 77243-580
Propan-2-yl 4-[5-methoxy-6-[(2-methyl-6-methylsulfonylpyridin-3-yl)amino]pyrimidin-4-yl]oxypiperidine-1-carboxylate 98%
Propan-2-yl 4-[5-methoxy-6-[(2-methyl-6-methylsulfonylpyridin-3-yl)amino]pyrimidin-4-yl]oxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₁H₂₉N₅O₆S
MW: 479.552 g/mol
CAS Number: 897732-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR