2,2'-Bithiophene-5-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Bithiophene-5-carboxaldehyde, 5-Formyl-2,2'-bithiophene

A710299-1G A710299-250MG A710299-5G
77267-646EA 28.84 USD
77267-646 77267-648 77267-650
2,2'-Bithiophene-5-carboxaldehyde 98%
2,2'-Bithiophene-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₆OS₂
MW: 194.27 g/mol
MDL Number: MFCD00052294
CAS Number: 3779-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR