3-Aminothiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol

A709756-25G A709756-5G
77280-896EA 62.83 USD
77280-896 77280-898
3-Aminothiophenol 98%
3-Aminothiophenol

Precaución: for Research Use Only

Formula: C₆H₇NS
MW: 125.19 g/mol
MDL Number: MFCD00007791
CAS Number: 22948-02-3
UN: 3267
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR