Methyl 1-methylpiperidine-2-carboxylate 98%

Supplier: Ambeed
Warning

A709097-100G A709097-25G A709097-5G
77243-552EA 73 USD
77243-552 77243-554 77243-556
Methyl 1-methylpiperidine-2-carboxylate 98%
Methyl 1-methylpiperidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MDL Number: MFCD09909832
CAS Number: 1690-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR