2,3-Benzofluorene 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Benzfluorene

A708811-100MG A708811-250MG
77255-220EA 38.11 USD
77255-220 77255-222
2,3-Benzofluorene 97%
2,3-Benzofluorene

Precaución: for Research Use Only

Formula: C₁₇H₁₂
MW: 216.28 g/mol
MDL Number: MFCD00003595
CAS Number: 243-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR