3,5-Bis(trifluoromethyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis(trifluoromethyl)pyrazole

A708395-1G A708395-250MG
77280-892EA 24.72 USD
77280-892 77280-894
3,5-Bis(trifluoromethyl)-1H-pyrazole 95%
3,5-Bis(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₂F₆N₂
MW: 204.07 g/mol
MDL Number: MFCD00153672
CAS Number: 14704-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR