3-Bromo-2,4,6-trimethylaniline 98%

Supplier: Ambeed
Warning

A708095-10G A708095-1G A708095-5G
77280-872EA 36 USD
77280-872 77280-874 77280-876
3-Bromo-2,4,6-trimethylaniline 98%
3-Bromo-2,4,6-trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MDL Number: MFCD00015476
CAS Number: 82842-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR