2,5-Dichloro-4-(3-nitrophenoxy)pyrimidine 97%

Supplier: Ambeed
Warning

A708006-100MG A708006-10G A708006-1G A708006-250MG A708006-5G
77267-626EA 108.15 USD
77267-626 77267-628 77267-630 77267-632 77267-634
2,5-Dichloro-4-(3-nitrophenoxy)pyrimidine 97%
2,5-Dichloro-4-(3-nitrophenoxy)pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₅Cl₂N₃O₃
MW: 286.07 g/mol
MDL Number: MFCD22124565
CAS Number: 76661-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR