2-Bromo-3,4-dimethoxybenzoic acid 97%

Supplier: Ambeed
Warning

A706917-100MG A706917-1G A706917-250MG
77267-620EA 77.25 USD
77267-620 77267-622 77267-624
2-Bromo-3,4-dimethoxybenzoic acid 97%
2-Bromo-3,4-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₄
MW: 261.06 g/mol
MDL Number: MFCD21340340
CAS Number: 71568-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR