4-[(tert-Butylamino)sulfonyl]benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-[(tert-Butylamino)sulphonyl]benzoic acid

A706795-1G A706795-250MG
77293-306EA 152.44 USD
77293-306 77293-308
4-[(tert-Butylamino)sulfonyl]benzoic acid 95%
4-[(tert-Butylamino)sulfonyl]benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₄S
MW: 257.306 g/mol
MDL Number: MFCD03001320
CAS Number: 99987-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR