1-Fluoro-3-nitrobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoronitrobenzene

A706758-10G A706758-25G A706758-5G A706758-100G
77243-534EA 8.24 USD
77243-534 77243-536 77243-538 77617-918
1-Fluoro-3-nitrobenzene 98%
1-Fluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₄FNO₂
MW: 141.1 g/mol
MDL Number: MFCD00007196
CAS Number: 402-67-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR