(3,3'-Difluoro-[2,2'-bithiophene]-5,5'-diyl)bis(trimethylstannane) 97%

Supplier: Ambeed
Danger

A705475-100MG A705475-1G A705475-250MG
77255-208EA 45 USD
77255-208 77255-210 77255-212
(3,3'-Difluoro-[2,2'-bithiophene]-5,5'-diyl)bis(trimethylstannane) 97%
(3,3'-Difluoro-[2,2'-bithiophene]-5,5'-diyl)bis(trimethylstannane)

Precaución: for Research Use Only

Formula: C₁₄H₂₀F₂S₂Sn₂
MDL Number: MFCD30543603
CAS Number: 1619967-09-7
UN: 3146
ADR: 6.1,Ⅰ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR