DBCO-NHS ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Succinimidyl 4-[(5-Aza-3,4:7,8-dibenzocyclooct-1-yne)-5-yl]-4-oxobutyrate, DBCO-NHS

A705235-100MG A705235-250MG A705235-50MG A705235-1G
77255-202EA 202.91 USD
77255-202 77255-204 77255-206 77552-140
DBCO-NHS ester 98%
DBCO-NHS ester

Precaución: for Research Use Only

Formula: C₂₃H₁₈N₂O₅
MW: 402.41 g/mol
MDL Number: MFCD24386367
CAS Number: 1353016-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR