H-Glu-Obzl.HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-Glu-Obzl·HCl

A705047-1G A705047-250MG
77243-512EA 12.36 USD
77243-512 77243-514
H-Glu-Obzl.HCl 97%
H-Glu-Obzl.HCl

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO₄
MW: 273.71 g/mol
MDL Number: MFCD18909979
CAS Number: 945651-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR