3-Ethoxy-2-cyclohexen-1-one 97%

Supplier: Ambeed
Warning

A705044-100G A705044-10G A705044-1G A705044-25G A705044-5G
77280-844EA 192 USD
77280-844 77280-846 77280-848 77280-850 77280-852
3-Ethoxy-2-cyclohexen-1-one 97%
3-Ethoxy-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00001580
CAS Number: 5323-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR