1-Phenylpiperidin-3-amine 97%

Supplier: Ambeed
Danger

A704530-1G A704530-250MG
77255-198EA 553.11 USD
77255-198 77255-200
1-Phenylpiperidin-3-amine 97%
1-Phenylpiperidin-3-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MW: 176.25 g/mol
MDL Number: MFCD11109783
CAS Number: 63921-21-1
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR