2-Mercaptopyridine-N-oxide 98%

Supplier: Ambeed
Warning

Synonyms: 2-Mercaptopyridine 1-oxide

A704218-100G A704218-25G A704218-5G
77267-604EA 311 USD
77267-604 77267-606 77267-608
2-Mercaptopyridine-N-oxide 98%
2-Mercaptopyridine N-oxide

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.17 g/mol
MDL Number: MFCD00006196
CAS Number: 1121-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR