3-Phenyl-1H-imidazo[4,5-b]pyridin-2(3H)-one 97%

Supplier: Ambeed
Warning

A703775-100MG A703775-1G A703775-250MG
77243-506EA 76.22 USD
77243-506 77243-508 77243-510
3-Phenyl-1H-imidazo[4,5-b]pyridin-2(3H)-one 97%
3-Phenyl-1H-imidazo[4,5-b]pyridin-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₂H₉N₃O
MW: 211.22 g/mol
MDL Number: MFCD01879476
CAS Number: 41010-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR