5-Iodo-1-methyl-1H-indazol-3-ol 97%

Supplier: Ambeed
Warning

A703669-100MG A703669-250MG
77243-502EA 52.53 USD
77243-502 77243-504
5-Iodo-1-methyl-1H-indazol-3-ol 97%
5-Iodo-1-methyl-1H-indazol-3-ol

Precaución: for Research Use Only

Formula: C₈H₇IN₂O
MW: 274.05 g/mol
MDL Number: MFCD28579521
CAS Number: 1226985-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR