1-(9H-Fluoren-9-yl)-3,12-dioxo-2,7,10,16,19-pentaoxa-4,13-diazahenicosan-21-oic acid 95%

Supplier: Ambeed
Warning

Synonyms: FmocNH-PEG2-CH2CONH-PEG2-CH2COOH

A703383-250MG A703383-5G
77255-188EA 60 USD
77255-188 77573-276
1-(9H-Fluoren-9-yl)-3,12-dioxo-2,7,10,16,19-pentaoxa-4,13-diazahenicosan-21-oic acid 95%
1-(9H-Fluoren-9-yl)-3,12-dioxo-2,7,10,16,19-pentaoxa-4,13-diazahenicosan-21-oic acid

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂O₉
MDL Number: MFCD09752868
CAS Number: 560088-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR