4-Nitrophenyl tetradecanoate 98%

Supplier: Ambeed
Warning

A703228-1G A703228-250MG A703228-5G
77293-296EA 25 USD
77293-296 77293-298 77293-300
4-Nitrophenyl tetradecanoate 98%
4-Nitrophenyl tetradecanoate

Precaución: for Research Use Only

Formula: C₂₀H₃₁NO₄
MDL Number: MFCD00047731
CAS Number: 14617-85-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR