SGC0946 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(3-((((2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)(isopropyl)amino)propyl)-3-(4-(tert-butyl)phenyl)urea
A702574-10MG
A702574-1MG
A702574-50MG
A702574-5MG
77243-494EA
140
USD
77243-494
77243-496
77243-498
77243-500
SGC0946 98%
SGC0946
Precaución: for Research Use Only
Formula:
C₂₈H₄₀BrN₇O₄ MW: 618.577 g/mol |
MDL Number:
MFCD26523136 CAS Number: 1561178-17-3 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...