rel-(1R,2R)-Cyclopropane-1,2-dicarboxylic acid 98%
Supplier: Ambeed
|
Danger
|
A702512-1G
A702512-250MG
A702512-5G
77243-488EA
35
USD
77243-488
77243-490
77243-492
rel-(1R,2R)-Cyclopropane-1,2-dicarboxylic acid 98%
rel-(1R,2R)-Cyclopropane-1,2-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₆O₄ |
MDL Number:
MFCD00034639 CAS Number: 696-75-3 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...