Zanubrutinib 97%

Supplier: Ambeed
Warning

Synonyms: (S)-7-(1-Acryloylpiperidin-4-yl)-2-(4-phenoxyphenyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxamide

A702104-10MG A702104-1MG A702104-50MG A702104-100MG
77267-598EA 119.48 USD
77267-598 77267-600 77749-442 77749-441
Zanubrutinib 97%
Zanubrutinib

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₅O₃
MW: 471.56 g/mol
MDL Number: MFCD31567461
CAS Number: 1691249-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR