4-Acetamido-3-ethoxynitrobenzene 95%

Supplier: Ambeed
Warning

Synonyms: N-(2-Ethoxy-4-nitrophenyl)acetamide

A701403-10G A701403-1G A701403-25G A701403-5G
77293-288EA 156.56 USD
77293-288 77293-290 77293-292 77293-294
4-Acetamido-3-ethoxynitrobenzene 95%
4-Acetamido-3-ethoxynitrobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₄
MW: 224.213 g/mol
MDL Number: MFCD00994990
CAS Number: 116496-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR