Methyl 2-(4-aminopiperidin-1-yl)nicotinate 97%

Supplier: Ambeed
Warning

A700891-100MG A700891-1G A700891-250MG
77243-482EA 118.45 USD
77243-482 77243-484 77243-486
Methyl 2-(4-aminopiperidin-1-yl)nicotinate 97%
Methyl 2-(4-aminopiperidin-1-yl)nicotinate

Precaución: for Research Use Only

Formula: C₁₂H₁₇N₃O₂
MW: 235.28 g/mol
CAS Number: 1185536-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR