2,6-Dichlorothiophenol 98%

Supplier: Ambeed
Danger

A700660-25G A700660-5G
77255-180EA 69 USD
77255-180 77255-182
2,6-Dichlorothiophenol 98%
2,6-Dichlorothiophenol

Precaución: for Research Use Only

Formula: C₆H₄Cl₂S
MDL Number: MFCD00004833
CAS Number: 24966-39-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR