3-(Benzo[d]thiazol-2-yl)-2-methylaniline 97%

Supplier: Ambeed
Warning

A700503-100MG A700503-1G A700503-250MG
77243-474EA 31 USD
77243-474 77243-476 77243-478
3-(Benzo[d]thiazol-2-yl)-2-methylaniline 97%
3-(Benzo[d]thiazol-2-yl)-2-methylaniline

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂S
MDL Number: MFCD00579124
CAS Number: 292644-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR